missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 96-Well, cellGrade-Treated, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Brand: BRAND™ 781968
263.85 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
- Easily binds serum components to for indirect adhesion of cells
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- DNA-, DNase-, RNase-free
- Endotoxins are <0.01 EU/mL
- Orange alphanumeric embossing for easy well identification
Order Info
Packs of 50 individually wrapped plates with lids
Specifications
ISO 11137, DIN EN ISO 10993 | |
With Lid | |
Carboxyl and Hydroxyl Groups | |
Polystyrene | |
Sterile | |
350 μL | |
Cultivation of adherent cell lines | |
1/Pk. |
Standard | |
8 x 12 | |
Black | |
96 | |
cellGrade | |
Flat Bottom (F-well) | |
50/Cs. |