Learn More
BRAND™ nano-cap™ Pipet Tips
Manufactured from high purity, DiHEMDA- and oleamide-free, virgin polypropylene. nano-cap™ Pipet Tips provide clean, hydrophobic surfaces, reduce sample retention, and increase accuracy and reproducibility.
Brand: BRAND™ 732002
54.80 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Precision-molded from high purity, DiHEMDA- and oleamide-free, virgin polypropylene.
- Clean, hydrophobic surfaces reduce sample retention, and increase accuracy and reproducibility.
- Available either in bulk packages or in convenient rack-and-stack formats
Specifications
0.1 to 20 μL | |
DiHEMDA- and oleamide-free | |
Nano-cap | |
20, 100μL |
Non-sterile | |
Bulk | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000013195 | Certificate of Analysis | 31/03/2025 | 732002 |
0000012867 | Certificate of Analysis | 17/02/2025 | 732002 |
0000012233 | Certificate of Analysis | 25/11/2024 | 732002 |
Promotions
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
10Hours
2Minutes
12Seconds
7 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
27Hours
2Minutes
12Seconds
7