Learn More
BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories
Allows the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.Dispensette™III, Dispensette™ Organic Remote Dispenser Parts and Accessories include threaded adapters, a table/shelf clamp and a support rod for the wall mounting unit.
Brand:  BRAND™ 704272
18.55 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10643202
  • 27.70€ / Each
Estimated Shipment: 30-05-2025
Description
  • Dispensette™ parts and accessories allow the Dispensette™ to be mounted on a wall, a ring stand, or on lab furniture.
  • Dispenses accurate volumes directly from drums and bulk refills.
  • A filter in the drum adapter minimizes risk of contaminating high-purity reagents.
  • A quick-release connector with integrated valves allows quick changing of the bulk container.
  • The remote dispensing system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
  • Maximum delivery height is approximately 1.2 m
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Table/Shelf Clamp for Wall Mounting Unit
Dispensette™III, Dispensette™ Organic
1 Ea.
Product Title
BRAND™ Dispensette™III, Dispensette™ Organic Remote Dispensing System Parts and Accessories > Table/Shelf Clamp for Dispensette wall mounting unit