Learn More
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Brand: BRAND™ 704396
10.34 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Polypropylene Construction
- For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
Specifications
Bottle Thread Adapter | |
Polypropylene | |
Dispensette™, seripettor™, Titrette™ and QuikSip™ | |
GL 45/33 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%