missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrdU Mouse anti-Human, Mouse, Rat, Clone: 1B10E12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66241-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody does not cross react with Thymidine.
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA.Specifications
BrdU | |
Monoclonal | |
1.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Mouse | |
150 μL | |
Primary | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1B10E12 | |
Unconjugated | |
5-bromo-2-deoxyuridine, BrDu, BrdUrd, Bromodeoxyuridine, BUdR | |
Protein A | |
RUO | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BrdU Mouse anti-Human, Mouse, Rat, Clone: 1B10E12, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction