Learn More
Mettler Toledo™ Bromide Ion Selective Electrode
Brand: Mettler Toledo™ 51340300
1140.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Solid State Membrane | |
0°C to 80°C | |
2 to 13 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 6
Product Content Correction
Product Title
Mettler Toledo™ Bromide Ion Selective Electrode > Membrane: Solid State