missing translation for 'onlineSavingsMsg'
Learn More

Bromosporine, Tocris Bioscience™

Product Code. 30001667
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Broad spectrum bromodomain inhibitor

Broad spectrum bromodomain inhibitor. Accelerates FRAP recovery of BRD4 and CREBBP in cells at a concentration of 1 μM.

Specifications

CAS 1619994-69-2
Molecular Formula C17H20N6O4S
Quantity 10 mg
Synonym bromosporine, bioomosporine, bromorsporine, d07alg, bromosporine hplc, carbamic acid, n-3-methyl-6-4-methyl-3-methylsulfonyl amino phenyl-1,2,4-triazolo 4,3-b pyridazin-8-yl-, ethyl ester, ethyl 3-methyl-6-4-methyl-3-methylsulfonamido phenyl-1,2,4 triazolo 4,3-b pyridazin-8-yl carbamate, ethyl n-6-3-methanesulfonamido-4-methylphenyl-3-methyl-1,2,4 triazolo 4,3-b pyridazin-8-yl carbamate, n-6-3-methanesulfonamido-4-methylphenyl-3-methyl-1,2,4 triazolo 4,3-b pyridazin-8-yl carbamate
InChI Key UYBRROMMFMPJAN-UHFFFAOYSA-N
SMILES CCOC(=O)NC1=CC(=NN2C1=NN=C2C)C3=CC(=C(C=C3)C)NS(=O)(=O)C
IUPAC Name ethyl N-[6-[3-(methanesulfonamido)-4-methylphenyl]-3-methyl-[1,2,4]triazolo[4,3-b]pyridazin-8-yl]carbamate
Molecular Weight (g/mol) 404.445
PubChem CID 72943187
Formula Weight 404.44
Percent Purity >98%
Chemical Name or Material Bromosporine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'