Learn More
Tocris Bioscience™ BTT 3033
Selective inhibitor of integrin α2β1
Brand:  Tocris Bioscience™ 4724/50
1046.19 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18733158
  • 1365.00€ / 50mg
Estimated Shipment: 11-12-2024
Description
BTT 3033 is a selective inhibitor of integrin α2β1 (EC50 = 130 nM for α2β1 binding to collagen I). Binds to the α2I domain. Exhibits selectivity for α2β1 over integrins α3β1, α4β1, α5β1 and αv. BTT 3033 inhibits platelet aggregation to collagen I coated capillaries under flow and inhibits binding of α2-expressing CHO cells to collagen I under shear stress conditions. Neurogenic and thromboxane A2-induced human prostate smooth muscle contraction are also inhibited by BTT 3033.
Specifications
Specifications
>98%
BTT 3033
BTT 3033
50 mg
465.5