Learn More
BUCHI Boiling Point Tubes and Capillaries
Boiling point tubes and capillaries for automated or manual determination according to the Siwoloboff method.
Brand: BUCHI 051890
This item is not returnable.
View return policy
Description
- High precision for reliable resutls
- Meets Siwoloboff and Eu Pharmacopeia standards
- Designed for BUCHI melting and boiling point instruments
- Compatible with other instruments
Specifications
Boiling Point Capillaries | |
Laboratory general purpose tubing | |
Borosilicate Glass 3.3 | |
Autoclavable |
0.8 mm | |
Boiling point apparatus | |
100/Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
BUCHI Boiling Point Tubes and Capillaries > Boiling point capillaries; Count: 100 pieces