missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffer A2 with Lyse Control
Lysis buffer for breaking down the cell membrane in order to purify plasmid DNA using NucleoSpin™ Plasmid kids.
Brand: Macherey-Nagel™ 740328.100
26.81 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
NucleoSpin™ 8/96 Plasmid Kit is used for isolation of plasmid DNA using either a flexible 8-well strip format for varying throughput or a proven 96-well plate format for high troughput.
- Time-saving parallel isolation of plasmid DNA
- Processing of up to 5 mL bacterial culture
- NucleoSpin™ 8 Filter Strips/96 Filter Plate for convenient filtration of bacterial lysates
- Processing under vacuum or by centrifugation
- Suitable for manual or automated processing under vacuum
- Innovative MN Wash Plate minimizes risk of cross-contamination
- Core Kit contains the basic content, focusing on automation platforms; additional accessories can be combined as needed
Specifications
Colorless | |
Dodium hydroxide diluted solution, Dodecyl sulfate, sodium salt | |
100 mL | |
Odorless | |
Liquid |
NucleoSpin Plasmid kits | |
>12 | |
1310-73-2 | |
Room Temperature (18°C to 25°C) | |
Plasmid DNA purification |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- Buffer A2 with Lyse Control (100 mL)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide, dodecyl sulphate