Learn More
Buffer Colour Coded Concentrated Solution pH10.00 (Borate) Blue, NIST Standard for pH measurement, Fisher Chemical™
23.10€ - 92.15€
Specifications
Physical Form Liquid
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10293862
View Documents
Fisher Chemical
J/2885C/05
1 Ampule This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
10615112
View Documents
Fisher Chemical
J/2885C/90
6 Ea. This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A