Learn More
Buffer Concentrated Solution BS pH 6.87 (Phosphate), Standard for pH measurement, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Specifications
Specifications
Physical Form | Liquid |
Quantity | 100 mL |
MDL Number | 282720 |
Packaging | Ampoule |
Safety and Handling
Product Identifier
- Buffer solution pH 6.87 BS formulated
Product Content Correction
Product Title
Buffer Concentrated Solution BS pH 6.87 (Phosphate), Standard for pH measurement, Fisher Chemical >