missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 4.00, NIST, Chem-Lab
Click to view available options
Quantity:
25 L
Packaging:
PE
Description
pH = 4.03 at 0°C
pH = 4.02 at 10°C
pH = 4.00 at 20°C
pH = 4.00 at 25°C
pH = 4.00 at 30°C
pH = 4.00 at 40°C
pH = 4.00 at 50°C
pH = 4.00 at 60°C
pH = 4.00 at 70°C
pH = 4.00 at 80°C
pH = 4.00 at 90°C
pH = 4.02 at 10°C
pH = 4.00 at 20°C
pH = 4.00 at 25°C
pH = 4.00 at 30°C
pH = 4.00 at 40°C
pH = 4.00 at 50°C
pH = 4.00 at 60°C
pH = 4.00 at 70°C
pH = 4.00 at 80°C
pH = 4.00 at 90°C
Specifications
Specifications
Density | 1.01g/mL |
pH | 4.00 |
Quantity | 25 L |
Formulation | Citrate, NaCl, NaOH |
Packaging | PE |
Traceability to NIST | NIST traceable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'