Learn More
Atago™ Bulbs
Use this bulb with Atago™ Illuminators for Abbe Refractometers.
Brand:  Atago™ RE-1170
Product Code. 11741579
  • POA / Pack of 5
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
  • Lamp for illuminator
  • 8V
  • 0.15A
  • Available in packs of five
Specifications
Specifications
Bulb
Abbe Refractometers NAR-1T, NAR-2T, NAR-3T, NAR-4T
Lamp for illuminator (8V, 0.15A)
5 Pk.
Product Title
Atago™ Bulbs > Electrical Requirements: 8V, 0.15A