Learn More
Rogo Sampaic™ Button Probe
Brand: Rogo Sampaic™ FMD006
This item is not returnable.
View return policy
Specifications
Button probe | |
Stainless Steel | |
Button | |
140 mm |
Product Content Correction
Product Title
Rogo Sampaic™ Button Probe > Length: 140mm