missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C-Reactive Protein/CRP Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
357.00€ - 756.00€
Specifications
Assay Range | 34.38 pg/mL |
---|---|
Assay Sensitivity | 34.38-2200 pg/mL |
Assay | Sandwich ELISA |
Conjugate | HRP |
Immunoassay Kit Format | Sandwich ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18273106
|
Novus Biologicals™
NBP2-79581-15PLATES |
15 Plates |
756.00€
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18245286
|
Novus Biologicals™
NBP2-79581-5PLATES |
5 Plates |
357.00€
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP.Specifications
34.38 pg/mL | |
Sandwich ELISA | |
Sandwich ELISA Kit | |
This solid phase sandwich ELISA utilizes an antibody specific for Mouse C-Reactive Protein/CRP. | |
Sandwich ELISA | |
1401 | |
Rabbit | |
RUO | |
Storage is content dependent. | |
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 2200 pg/mL is recommended. |
34.38-2200 pg/mL | |
HRP | |
Antibody Pairs | |
Mouse | |
C-reactive protein, C-reactive protein, pentraxin-related, MGC88244, pentraxin 1, PTX1MGC149895 | |
CRP | |
Cancer, Cytokine Research, Immunology, Innate Immunity | |
C-Reactive Protein/CRP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ C-Reactive Protein/CRP Antibody Pair [HRP]