missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals C-telopeptide

For cellular and molecular biology applications

91.05€ - 169.00€

Specifications

Purification Method HPLC
Common Name C-terminal telopeptide
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 868.89
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15438009
View Documents
Thermo Scientific Chemicals
J66595.!D
0.5 mg
91.05€
0.50mg
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
15448009
View Documents
Thermo Scientific Chemicals
J66595.M¬
1 mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

HPLC
-30°C to -10°C
868.89
RUO - research use only
C-terminal telopeptide
Solid
C-telopeptide
Glu-Lys-Ala-His-Asp-Gly-Gly-Arg
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.