Learn More
Cadmium Solution for ICP, 100mg/L, HNO3 Matrix, Fisher Chemical™ Product Code.: 12997253
Cadmium Solution for ICP, 100mg/L, HNO3 Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12997253 Shop All Fisher Chemical Products
100 mL
Click to view available options
Quantity:
100 mL
Product Code. 12997253
Brand: Fisher Chemical S 410 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains HNO3
Concentration 100 mg/L
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1111517 Certificate TypeCertificate of Analysis Date22/05/2025 Catalog NumberS 410 100
Lot Number1071533 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS 410 100
Lot Number1049318 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberS 410 100
Lot Number1000227 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberS 410 100
Product Content Correction
Product Title
Cadmium Solution for ICP, 100mg/L, HNO3 Matrix, Fisher Chemical™ >