Learn More
R&D Systems™ Calcein AM
R&D Systems™ Calcein AM provides a simple, rapid and accurate method to measure cell viability and/or cytotoxicity.
Brand: R&D Systems™ 4892-010-01
Additional Details : Weight : 0.09700kg
Description
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
- Do not use past expiration date.
- The hydrolysis of Calcein AM by intracellular esterases produces calcein (structure B), a hydrophilic, strongly fluorescent compound that is well-retained in the cell cytoplasm.
- Cells grown in black-walled plates can be stained and quantified in less than two hours.
- Non-fluorescent, hydrophilic compound that easily permeates intact, live cells
Specifications
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
50 μg |