Learn More
Endecotts™ Sieve Calibration Standards
Use for calibration of the woven wire mesh sieves. All reference samples supplied by Endecotts are provided with a Certificate of Calibration and are fully traceable to National Standards
Brand:  Endecotts™ ZSICSA-2.36
 View more versions of this product
Product Code. 11904465
  • 441.00€ / Pack of 2
Estimated Shipment: 25-02-2025
Description
  • Calibrates sieves to an accuracy of ±1μm
  • Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture.
  • Traceable to the United Kingdom National Physical Laboratory
  • Standards include a calibration chart.
Specifications
Specifications
Calibration Ball Mesh
Sieve
2.36 mm
Soda-lime glass
2/Pk.