Learn More
Carl Zeiss™ Camera Adapters
C-mount camera adapter for Primovert
173.00€ - 1025.00€
Product Code | Brand | Description | Magnification Power | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Magnification Power | Price | Quantity & Availability | |||||
11875873
|
Carl Zeiss™
426116-0000-000 |
SLR | 2.5X |
928.00€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
11856432
|
Carl Zeiss™
426115-0000-000 |
T2-T2 DSLR | 1.6X |
892.00€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
11895440
|
Carl Zeiss™
426112-0000-000 |
60N-C 0.75 in. | 0.5x |
990.00€
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
10337143
|
Carl Zeiss™
415500-1810-000 |
P95-C 0.75 in. | 0.65x |
385.00€
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
10397183
|
Carl Zeiss™
415500-1811-000 |
P95-C 0.50 in. | 0.5x |
376.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
10875741
|
Carl Zeiss™
426113-0000-000 |
60N-C 0.75 in. | 0.63x |
1025.00€
Each |
Estimated Shipment: 28-04-2025
Log in to see stock. |
|||||
15150705
|
Carl Zeiss™
426114-0000-000 |
60N-C 1.0 in. | 1.0X |
173.00€
1 unit |
Estimated Shipment: 26-05-2025
Log in to see stock. |
|||||
Description
- Requires Primovert
- With a phototube C mount camera adapter
- For Primovert
Specifications
Camera Adapter | |
1 Ea. |
Product Title
Carl Zeiss™ Camera Adapters