Learn More
Thermo Scientific Chemicals cAMP Dependent PK Inhibitor, PKI (14-24), Amide
For cellular and molecular biology applications
58.12€ - 102.51€
Specifications
Purification Method | HPLC |
---|---|
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 1251.37 |
Product Type | cAMP Dependent PK Inhibitor, PKI (14-24), amide |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
cAMP Dependent PK Inhibitor, PKI (14-24), amide | |
Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2 |
-30°C to -10°C | |
1251.37 | |
RUO - research use only |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.