missing translation for 'onlineSavingsMsg'
Learn More
(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals Product Code.: 11458960

(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals

Product Code. 11458960
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11458960

Brand: Thermo Scientific Alfa Aesar A14967.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Camphorquinone derivative (i.e. carboxylated camphorquinone) is used as visible-light photoinitiator in biomedical applications. It is also used in dentistry and is often used in conjunction with hydrogen donating amine.

Solubility
Soluble in ethanol, ethyl ether and benzene.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10373-78-1
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00064160
InChI Key VNQXSTWCDUXYEZ-UHFFFAOYSA-N
Synonym camphorquinone, dl-camphorquinone, camphoroquinone, 2,3-bornanedione, camphor quinone, camphoquinone, 1,7,7-trimethylbicyclo 2.2.1 heptane-2,3-dione, bornane-2,3-dione, bicyclo 2.2.1 heptane-2,3-dione, 1,7,7-trimethyl, +/--camphorquinone
PubChem CID 25208
ChEBI CHEBI:34607
IUPAC Name 4,7,7-trimethylbicyclo[2.2.1]heptane-2,3-dione
SMILES CC1(C2CCC1(C(=O)C2=O)C)C

Specifications

Melting Point 198°C to 202°C
Quantity 100 g
Beilstein 1909463
Solubility Information Soluble in ethanol,ethyl ether and benzene.
Formula Weight 166.22
Percent Purity 99%
Chemical Name or Material (+/-)-Camphorquinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249559 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog NumberA14967.06,A14967.22,A14967.14
Lot Number10245865 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14967.22
Lot Number10242875 Certificate TypeCertificate of Analysis Date16/08/2022 Catalog NumberA14967.06,A14967.22,A14967.14
Lot Number10239768 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14967.06,A14967.22,A14967.14
Lot Number10217209 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14967.06,A14967.22,A14967.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+/-)-Camphorquinone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.