Learn More
Hirschmann™ Capillary Tubes, Both Ends Open Product Code.: 13220490
  • ONLINE EXCLUSIVE PRICE
    128.00€
    123.00€ / Pack of 1000
    Save 5.00€
    3% Off
Hirschmann™ Capillary Tubes, Both Ends Open
Product Code. 13220490
100 mm, 1.55 mm, Pack of 1000
missing translation for 'orderingAttributeHoverText'
Length (Metric) Tube:
60 mm
70 mm
75 mm
80 mm
90 mm
100 mm
120 mm
150 mm
Diameter (Metric) Inner:
0.95 mm
1.15 mm
1.30 mm
1.55 mm
missing translation for 'unitSize'
Pack of 1000
Product Code. 13220490
Brand: Hirschmann™ 9202310
  • ONLINE EXCLUSIVE PRICE
    128.00€
    123.00€ / Pack of 1000
    Save 5.00€
    3% Off
Excellent for working with small samples in the laboratory. Can be utilized for a wide range of scientific and medical applications.
  • Both ends open
  • Available in a wide range of diameters/lengths
Specifications
Product Type Capillary Tubes, Both Ends Open
Diameter (Metric) Inner 1.55 mm
Length (Metric) Tube 100 mm
Quantity 4 x 250/Pk.
Product Content Correction
Product Title
Hirschmann™ Capillary Tubes, Both Ends Open >