Learn More
Hirschmann™ Capillary Tubes for Melting Point Determination, One End Closed
Excellent for melting point analysis in the laboratory
Brand:  Hirschmann™ 9211570
 View more versions of this product
Product Code. 10009760
  • 84.05€ / Pack of 1000
Estimated Shipment: 19-02-2025
Description
  • Capillary tubes
  • One end closed
  • Available in a wide range of diameters/lengths
Specifications
Specifications
Melting Point Determination Tube, One End Closed
1.15 mm
70 mm
10 x 100/Pk.