missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate

Thermo Scientific™ CaptureSelect™ Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT).

514.00€ - 1802.00€

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Label Type Biotin and Analogs
Product Line CaptureSelect™
Target Species Human
Ligand Type Antibody
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18446638
View Documents Promotion Details
Thermo Scientific™
7102872100
100 μg
514.00€
100µg
In Stock
to see stock.
Add to basket
 
18496778
View Documents
Thermo Scientific™
7102872500
500 μg
1802.00€
500µg
In Stock
to see stock.
Add to basket
 
Description

Description

CaptureSelect Biotin Anti-AAT Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity and selectivity to human alpha-1 antitrypsin (AAT). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize native and recombinant human AAT.

Applications for CaptureSelect Biotin Anti-AAT Conjugate include ELISA, immunoprecipitation with streptavidin-coupled DynaBeads, Gyrolab™-based immunoassays, and label-free detection using platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).

Animal origin statement

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Specifications

Specifications

Store in freezer (-5°C to -30°C).
CaptureSelect™
Antibody
Biotin and Analogs
Human
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.