Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAV9 Conjugate
CaptureSelect™ Biotin Anti-AAV9 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to adeno-associated virus particles of serotype 9 (AAV9).
514.00€ - 1802.00€
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
Product Line | CaptureSelect™ |
Ligand Type | Antibody |
Shipping Condition | Dry Ice |
Description
CaptureSelect™ Biotin Anti-AAV9 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that specifically binds with high affinity to adeno-associated virus particles of serotype 9 (AAV9). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AAV9 particles.
Applications for CaptureSelect Biotin Anti-AAV9 Conjugate include ELISA, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Specifications
Store in freezer (-5°C to -30°C). | |
CaptureSelect™ | |
Antibody | |
Dry Ice |