Learn More
Carbon, activated, -4+8 mesh, Thermo Scientific Chemicals Product Code.: 11324788
Carbon, activated, -4+8 mesh, Thermo Scientific Chemicals
Product Code. 11324788
100 g
Click to view available options
Quantity:
100 g
500 g
2 kg
Product Code. 11324788
Brand: Thermo Scientific Alfa Aesar 043118.22
Request Bulk or Custom Format
CAS: 1333-86-4 | C | 12.01 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Specifications
Mesh Size −4+8 Mesh
Quantity 100 g
Chemical Name or Material Carbon, Activated
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ08J017 Certificate TypeCertificate of Analysis Date04/01/2024 Catalog Number043118.36,043118.22,043118.A3
Lot NumberY13J028 Certificate TypeCertificate of Analysis Date28/12/2023 Catalog Number043118.36,043118.22,043118.A3
Lot NumberW07J024 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number043118.36,043118.22,043118.A3
Lot NumberR31J051 Certificate TypeCertificate of Analysis Date21/06/2023 Catalog Number043118.36,043118,043118.22,043118.A3
Lot NumberW11G036 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043118.36,043142.AK,043118.22,043118.A3
Product Title
Carbon, activated, -4+8 mesh, Thermo Scientific Chemicals >