Learn More
Cytiva Carbon Capsule - 100 g Activated Carbon
Carbon capsule not approved for use in medical, clinical, surgical or other patient applications.
Brand:  Cytiva 12011
132.21 EUR valid until 2025-06-21
Use promo code "24097" to get your promotional price.
Product Code. 17144211
  • 194.00€ / Each
Outlet Corner
Use promo code 24097 at checkout
View Available Promotions
Edge
Description
Description
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Activated Carbon
Non-sterile
70,000 m2
3/8 in. FNPT or 1/2 in. hose barb
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Use code 24097 to receive this promotion.
    Promo Code: 24097
    Expires2025-06-16
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 19/05/2025 and 16/06/2025 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions Learn More
Product Content Correction
Product Title
Cytiva Carbon Capsule - 100 g Activated Carbon > Non-sterile