Learn More
Swann-Morton™ Carbon Steel Individual Peel Pack, Non-Sterile Scalpel Blades, No. 10
Click to view available options
Quantity:
100/Box
Unit Size:
Pack of 100
Description
- Curved cutting edge.
- Traditional blade shape.
- Used generally for making varying sizes of incision in skin and muscle.
- Often used in more specialized surgeries such as harvesting the radial artery during a coronary artery bypass operation, opening the bronchus during thoracic surgery, uterine incisions for a C-section and for inguinal hernia repair.
- Fits handles 3, 3L, 3 graduated, 5B, 7, 9, B3.
Specifications
Specifications
| Sterility | Non-sterile |
| Quantity | 100/Box |
| Packaging Type | Individually Packaged |