Learn More
Novus Biologicals™ Carbonic Anhydrase X/CA10 Overexpression Lysate
Brand:  Novus Biologicals™ NBP2-09282
Product Code. 18051539
  • 700.00€ / 0.10mg
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Carbonic Anhydrase X/CA10 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_1763. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens CA10, mRNA.
Native