Learn More
Thermo Scientific Chemicals Casein, pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Casein contains a mixture of related phosphoproteins present in bovine milk and cheese. It contains all of the common amino acids and is rich in the essential ones.
Applications
• Casein is a source of amino acids, carbohydrates, and two inorganic elements namely calcium and phosphorus
• This compound can be used in vivo as a solid food sample and in vitro digestion experiments with rodents
• It can increase the photostability of quantum dots and decrease nonspecific binding in immunofluorescence experiments
• Casein, in biochemical experiments, can be used to prepare P407-casein hydrogels and to study the mechanical effects of the addition of casein to P407
Literature References
Tundisi, L.; Yang, R.; Borelli, L.; Alves, T.; Mehta, M.; Chaud, M.; Mazzola, P.; Kohane, D. Enhancement of the mechanical and drug-releasing properties of poloxamer 407 hydrogels with casein. Pharm Res. 2021, 38 (3), 515–522.
Irshad, I.; Kanekanian, A.; Peters, A.; Masud, T. Antioxidant activity of bioactive peptides derived from bovine casein hydrolysate fractions. J. Food Sci. Technol. 2015, 52 (1), 231-239.
Sharma, A. K.; Sawhney, I. K. Modelling moisture sorption characteristics in dried acid casein using connectionist paradigm vis-Ă -vis classical methods. J. Food Sci. Technol. 2015, 52 (1), 151-160.

Specifications
Specifications
Melting Point | 280.0°C |
Packaging | Fibre drum |
Quantity | 25 kg |
Merck Index | 15,1883 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in conc. hcl with light violet color,soluble in non-polar organic solvents |
Grade | Pure |
Chemical Name or Material | Casein, tech. |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.