Learn More
3M™ Cashew Protein ELISA Kit, E96CHW
Intended for the detection of cashew proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients, and processed food products.
Brand:  3M™
Encompass
Product Code. 17263794
  • 793.53€ / 1 set
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to detect both processed and unprocessed target allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
  • A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Specifications
Test
96 Wells
50 Minutes