Learn More
3M™ Cashew Protein ELISA Kit, E96CHW
Intended for the detection of cashew proteins in clean-in-place water (CIP) final rinse water, environmental swab samples, food ingredients, and processed food products.
Brand: 3M™
This item is not returnable.
View return policy
Description
- Designed to detect both processed and unprocessed target allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples.
- A universal extraction protocol streamlines testing for multiple analytes, saving you reagents and valuable time.
Specifications
Test | |
96 Wells | |
50 Minutes |