Learn More
Thermo Scientific Chemicals Caspase-3 Substrate V, fluorogenic
For cellular and molecular biology applications
124.62€ - 229.00€
Specifications
Color | White to off-white |
---|---|
Form | Lyophilized |
Substrate Type | Caspase Substrate |
Molecular Weight (g/mol) | 1360.38 |
Storage Requirements | -30° to -10°C |
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Substrate Quantity | Price | Quantity & Availability | |||||
15488249
|
Thermo Scientific Chemicals
J66767.M^ |
1mg | N/A | ||||||
15478249
|
Thermo Scientific Chemicals
J66767.!D |
0.5mg |
124.62€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White to off-white | |
Caspase Substrate | |
-30° to -10°C |
Lyophilized | |
1360.38 |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.