Learn More
Thermo Scientific Chemicals Caspase-7 Substrate I, fluorogenic
For cellular and molecular biology applications
130.00€ - 248.00€
Specifications
Color White to off-white
Form Lyophilized
Substrate Type Caspase Substrate
Molecular Weight (g/mol) 1327.31
Storage Requirements -30° to -10°C
View More Specs
Products 2
Product Code Brand Substrate Quantity Price Quantity & Availability  
Product Code Brand Substrate Quantity Price Quantity & Availability  
15497899
View Documents
Thermo Scientific Chemicals
J66513.!D
0.5mg
130.00€
0.50mg
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
15407909
View Documents
Thermo Scientific Chemicals
J66513.M¬
1mg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
White to off-white
Caspase Substrate
-30° to -10°C
Lyophilized
1327.31
Videos
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C