missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAT Mouse anti-Human, Clone: OTI1B8, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502564S

This item is not returnable.
View return policy
Description
CAT Monoclonal antibody specifically detects CAT in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
CAT | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P04040 | |
Mouse | |
Affinity chromatography | |
RUO | |
847 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B8 | |
Unconjugated | |
Cat | |
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
Full length human recombinant protein of human CAT produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
CAT Mouse anti-Human, Clone: OTI1B8, TrueMAB™, Origene
Spot an opportunity for improvement?Share a Content Correction