missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Caution Labels

Brand:  Brady™ 225258

 View more versions of this product

Product Code. 11746835

  • 26.90€ / 1 pack
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

10 Pk.
TAG-OBLIGATION GEN.-75X160MM
Polyester
Lockout Tagout
160 mm
3 in.
-40°C to +100°C
2.95 in.
7 mm
Tag
Blue on White
Obligation Gen.
6.3 in.
75 mm
0.249 mm
-40°F to +212°F
75 mm
Lockout Tagout Tags
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.