missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 (IL-2 Receptor α) Mouse anti-Human, BUV563, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
499.00€ - 790.00€
Specifications
Antigen | CD25 (IL-2 Receptor α) |
---|---|
Clone | 2A3 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 563 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17207974
|
BD Biosciences
612919 |
25 Tests |
499.00€
25 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
17297964
|
BD Biosciences
612918 |
100 Tests |
790.00€
100 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Human samples. It is validated for Flow Cytometry.Specifications
CD25 (IL-2 Receptor α) | |
Flow Cytometry | |
Brilliant Ultraviolet 563 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
2A3 | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD25 (IL-2 Receptor α) |