missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25 Mouse anti-Human, Brilliant Ultraviolet 737, Clone: 2A3, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 612807
194.37 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry.
Specifications
CD25 | |
Monoclonal | |
Brilliant Ultraviolet 737 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
2A3 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human Phytohemagglutinin-activated T Cells | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction