missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD27 Mouse anti-Human, Alexa Fluor 647, Clone: O323, BD Biosciences™
Mouse Monoclonal Antibody
327.00€ - 890.00€
Specifications
Antigen | CD27 |
---|---|
Clone | O323 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 647 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17207814
|
BD Biosciences
567026 |
25 Tests |
327.00€
25 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
17297804
|
BD Biosciences
567025 |
100 Tests |
890.00€
100 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow cytometrySpecifications
CD27 | |
Flow Cytometry | |
Alexa Fluor 647 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Not Reported | |
Primary |
O323 | |
Monoclonal | |
Mouse | |
Human | |
CD27 antigen; TNFRSF7; S152; T14; Tp55 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. |