missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD279 (PD-1) Mouse anti-Human, Brilliant Ultraviolet 737, Clone: EH12.1 (also known as EH12), BD Horizon™
Mouse Monoclonal Antibody
388.00€ - 765.00€
Specifications
Antigen | CD279 (PD-1) |
---|---|
Clone | EH12.1 (also known as EH12) |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 737 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17217894
|
BD Biosciences
612792 |
25 Tests |
388.00€
25 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
17207894
|
BD Biosciences
612791 |
100 Tests |
765.00€
100 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
CD279 (PD-1) Monoclonal antibody specifically detects CD279 (PD-1) in Human samples. It is validated for Flow cytometry.Specifications
CD279 (PD-1) | |
Flow Cytometry | |
Brilliant Ultraviolet 737 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
EH12.1 (also known as EH12) | |
Monoclonal | |
Mouse | |
Human | |
Human PD-1 Recombinant Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |