Learn More
CD3 Mouse anti-Human, BB515, Clone: SK7, Secondary Antibody, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 564560
269.52 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
CD3 | |
Monoclonal | |
Brilliant Blue 515 (BB515) | |
100 Tests | |
Secondary | |
Human | |
IgG1 κ |
Flow Cytometry | |
SK7 (also known as Leu-4) | |
Mouse | |
RUO | |
CD3 | |
Refrigerate (2 to 8°C) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.