missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 Mouse anti-Human, NA/LE, Clone: OKT3, BD Biosciences™
Mouse Monoclonal Antibody
210.00€ - 1025.00€
Specifications
Antigen | CD3 |
---|---|
Clone | OKT3 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry |
Classification | Low Endotoxin/Azide-free (LE/AF) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17297824
|
BD Biosciences
567107 |
0.1 mg |
210.00€
100µg |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
17207844
|
BD Biosciences
567118 |
5.0 mg |
1025.00€
5mg |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry, ActivationSpecifications
CD3 | |
1.0 mg/mL | |
Low Endotoxin/Azide-free (LE/AF) | |
RUO | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. |
OKT3 | |
Flow Cytometry | |
Mouse | |
Human | |
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
IgG2a κ | |
Purified |