Learn More
CD33 Mouse, anti-Human, R718, Clone: WM53, BD Horizon™
Mouse Monoclonal Antibody
256.00€ - 550.00€
Specifications
Antigen | CD33 |
---|---|
Clone | WM53 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17217764
|
BD Biosciences
566972 |
25 Tests |
256.00€
25 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
17207764
|
BD Biosciences
566971 |
100 Tests |
550.00€
100 tests |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
The WM53 monoclonal antibody specifically recognizes CD33 which is also known as Sialic acid-binding Ig-like lectin 3 (Siglec-3) or gp67. CD33 is a 67 kDa type I transmembrane glycoprotein that is variably expressed on myeloid progenitors, monocytes, macrophages, dendritic cells, neutrophils, basophils, mast cells, and on some activated T cells and NK cells. Normal lymphocytes, platelets, erythrocytes and pluripotent hematopoietic stem cells do not express the CD33 antigen. This glycoprotein reportedly functions as a sialic acid-dependent cell adhesion molecule and this function can be modulated by endogenous sialoglycoconjugates when CD33 is expressed on the membrane.
Specifications
CD33 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
WM53 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |