missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD45 Mouse, anti-Human, R718, Clone: HI30, BD Horizon™
Mouse Monoclonal Antibody
202.00€ - 372.00€
Specifications
Antigen | CD45 |
---|---|
Clone | HI30 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | R718 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17217754
|
BD Biosciences
566962 |
25 Tests |
202.00€
25 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
17207754
|
BD Biosciences
566961 |
100 Tests |
372.00€
100 tests |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||||
Description
The HI30 monoclonal antibody specifically binds to the 180, 190, 205, 220 kDa protein isoforms of CD45. CD45 is encoded by the PTPRC (Protein tyrosine phosphatase receptor type C) gene. CD45, also known as the leukocyte common antigen (LCA), is a member of the protein tyrosine phosphatase (PTP) family. It is present on all human leukocytes including lymphocytes, monocytes, granulocytes, eosinophils, and thymocytes. CD45 is absent from circulating erythrocytes, platelets, or mature erythroid cells of bone marrow and non-hemopoietic tissues.
Specifications
CD45 | |
Flow Cytometry | |
R718 | |
Mouse | |
Human | |
Refrigerate Upon Arrival |
HI30 | |
Monoclonal | |
Liquid | |
RUO | |
Primary |