missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD95 (Fas) Mouse anti-Human, Baboon, Cynomolgus, Rhesus, BB700, Clone: DX2, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566543
This item is not returnable.
View return policy
Description
CD95 (Fas) Monoclonal antibody specifically detects CD95 (Fas) in Human,Baboon,Cynomolgus,Rhesus samples. It is validated for Flow Cytometry.Specifications
CD95 (Fas) | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
25 Tests | |
Primary | |
Human, Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
DX2 | |
Mouse | |
RUO | |
CD95 (Fas) | |
Refrigerate (2 to 8°C) |