missing translation for 'onlineSavingsMsg'
Learn More

Ceftizoxime, Thermo Scientific Chemicals

Product Code. 16504474
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68401-81-0
Molecular Formula C13H13N5O5S2
Molecular Weight (g/mol) 383.40
MDL Number MFCD00072000
InChI Key NNULBSISHYWZJU-LLKWHZGFSA-N
IUPAC Name sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-5-yl)-2-(methoxyimino)acetamido]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [H][C@]12SCC=C(N1C(=O)[C@H]2NC(=O)C(=N/OC)\C1=CSC(N)=N1)C(O)=O

Specifications

Melting Point 260.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 383.4
Chemical Name or Material Ceftizoxime

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ceftizoxime, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.