missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ 384-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:White; Sterility:Non-sterile; Lid:With Lid
Brand: Greiner Bio-One 781945
This item is not returnable.
View return policy
Description
- Improves adhesion of different cell types to polystyrene surfaces
- Reduced-serum or serum-free cultivation
- Increase in isolation and cultivation efficiency
- Ready-to-use products, time-saving
- Consistent quality
Specifications
Standard | |
16 x 24 | |
White | |
384 | |
Non-sterile | |
131 μL | |
4800 x g (swinging bucket rotor) | |
5 | |
Room Temperature |
With Lid | |
Not Autoclavable | |
Polystyrene | |
15 to 110 μL | |
Poly-D-Lysine | |
Flat Bottom (F-well) | |
20 | |
20/Cs. |