missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Flasks helps in promotion of cell adhesion and proliferation
Brand: Greiner Bio-One 658940
This item is not returnable.
View return policy
Description
Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.- Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Red | |
250 mL | |
Canted | |
5 | |
Yes | |
75 cm2 |
HDPE | |
Polystyrene | |
50 | |
Collagen Type I Protein | |
Filter Screw Cap | |
15 to 38 mL |