Learn More
Greiner Bio-One CELLMASTER Polystyrene 2.5X Roller Bottles - Short Form Product Code.: 10456342
Greiner Bio-One CELLMASTER Polystyrene 2.5X Roller Bottles - Short Form
Product Code. 10456342
24, 24
Click to view available options
No. per Pack:
24
No. per Case:
24
Product Code. 10456342
Brand: Greiner Bio-One 681075
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
For use in cell culture. Greiner Bio-One CELLMASTER Polystyrene 2.5X Roller Bottles - Short Form renders cell adhesion through physical surface treatment.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic, non-cytotoxic
  • Seamless blow-moulding technique, rules out leaking seams
  • USP Class VI end product testing certified
  • Safety screw cap guarantees tightly closed, co
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Ribbed
Capacity (Metric) 2100 mL
Color Clear
Diameter (English) 4.8 in.
For Use With (Application) Cell cuture
Material Polystyrene
No. per Case 24
No. per Pack 24
Sterility Sterile
Cell Growth Area 2125cm2
Certified USP Class VI end product testing
Format Short form
Surface Treatment TC Treated
Height (Metric) 271 mm
Product Type Roller Bottle
Diameter (Metric) 122 mm
Closure Type Screw Cap
Product Title
Greiner Bio-One CELLMASTER Polystyrene 2.5X Roller Bottles - Short Form >